Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601945 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SFSWAP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SFSWAP - splicing factor SWAP homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261411.1 | Intron | NP_001248340.1 | ||||
NM_004592.3 | Intron | NP_004583.2 | ||||
XM_011538653.1 | Intron | XP_011536955.1 | ||||
XM_011538654.2 | Intron | XP_011536956.1 | ||||
XM_011538655.2 | Intron | XP_011536957.1 | ||||
XM_017019798.1 | Intron | XP_016875287.1 | ||||
XM_017019799.1 | Intron | XP_016875288.1 |
Set Membership: |
HapMap |