Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609235 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRSK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
BRSK1 - BR serine/threonine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032430.1 | Intron | NP_115806.1 | ||||
XM_005259327.3 | Intron | XP_005259384.1 | ||||
XM_011527395.2 | Intron | XP_011525697.2 |
TMEM150B - transmembrane protein 150B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085488.2 | Intron | NP_001078957.1 | ||||
NM_001282011.1 | Intron | NP_001268940.1 | ||||
XM_005258812.3 | Intron | XP_005258869.1 | ||||
XM_011526850.2 | Intron | XP_011525152.1 | ||||
XM_011526851.2 | Intron | XP_011525153.1 | ||||
XM_011526852.2 | Intron | XP_011525154.1 | ||||
XM_011526853.2 | Intron | XP_011525155.1 | ||||
XM_011526854.2 | Intron | XP_011525156.1 | ||||
XM_011526855.2 | Intron | XP_011525157.1 | ||||
XM_011526856.2 | Intron | XP_011525158.1 | ||||
XM_011526857.2 | Intron | XP_011525159.1 | ||||
XM_017026667.1 | Intron | XP_016882156.1 | ||||
XM_017026668.1 | Intron | XP_016882157.1 |
Set Membership: |
HapMap Validated |