Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAGAGCTACTTTTCCCATATAAC[C/T]TAAACAATTTTCCCAAATAATAGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608767 | ||||||||||||||||||||
Literature Links: |
FEM1C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FEM1C - fem-1 homolog C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020177.2 | 5478 | UTR 3 | NP_064562.1 | |||
XM_005272035.4 | 5478 | UTR 3 | XP_005272092.1 | |||
XM_017009647.1 | 5478 | UTR 3 | XP_016865136.1 |