Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608828 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf22 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C5orf22 - chromosome 5 open reading frame 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018356.2 | Intron | NP_060826.2 | ||||
XM_006714479.2 | Intron | XP_006714542.1 | ||||
XM_006714480.3 | Intron | XP_006714543.1 | ||||
XM_011514062.2 | Intron | XP_011512364.1 | ||||
XM_017009607.1 | Intron | XP_016865096.1 | ||||
XM_017009608.1 | Intron | XP_016865097.1 | ||||
XM_017009609.1 | Intron | XP_016865098.1 | ||||
XM_017009610.1 | Intron | XP_016865099.1 | ||||
XM_017009611.1 | Intron | XP_016865100.1 | ||||
XM_017009612.1 | Intron | XP_016865101.1 | ||||
XM_017009613.1 | Intron | XP_016865102.1 | ||||
XM_017009614.1 | Intron | XP_016865103.1 | ||||
XM_017009615.1 | Intron | XP_016865104.1 | ||||
XM_017009616.1 | Intron | XP_016865105.1 |
DROSHA - drosha ribonuclease III | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100412.1 | Intron | NP_001093882.1 | ||||
NM_013235.4 | Intron | NP_037367.3 | ||||
XM_005248291.3 | Intron | XP_005248348.1 | ||||
XM_005248292.3 | Intron | XP_005248349.1 | ||||
XM_005248293.3 | Intron | XP_005248350.1 | ||||
XM_005248294.3 | Intron | XP_005248351.1 | ||||
XM_011514033.2 | Intron | XP_011512335.1 | ||||
XM_017009399.1 | Intron | XP_016864888.1 | ||||
XM_017009400.1 | Intron | XP_016864889.1 | ||||
XM_017009401.1 | Intron | XP_016864890.1 | ||||
XM_017009402.1 | Intron | XP_016864891.1 |
Set Membership: |
HapMap Validated |