Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611482 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD37 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANKRD37 - ankyrin repeat domain 37 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C4orf47 - chromosome 4 open reading frame 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114357.1 | Intron | NP_001107829.1 | ||||
XM_005263019.3 | Intron | XP_005263076.1 | ||||
XM_011531989.2 | Intron | XP_011530291.1 | ||||
XM_017008236.1 | Intron | XP_016863725.1 | ||||
XM_017008237.1 | Intron | XP_016863726.1 | ||||
XM_017008238.1 | Intron | XP_016863727.1 | ||||
XM_017008239.1 | Intron | XP_016863728.1 | ||||
XM_017008240.1 | Intron | XP_016863729.1 | ||||
XM_017008241.1 | Intron | XP_016863730.1 | ||||
XM_017008242.1 | Intron | XP_016863731.1 |
UFSP2 - UFM1 specific peptidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018359.3 | Intron | NP_060829.2 |