Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
56 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147264 MIM: 600172 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INPP5B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INPP5B - inositol polyphosphate-5-phosphatase B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297434.1 | 2581 | UTR 3 | NP_001284363.1 | |||
NM_005540.2 | 2581 | UTR 3 | NP_005531.2 | |||
XM_006710621.3 | 2581 | Intron | XP_006710684.2 | |||
XM_011541392.2 | 2581 | UTR 3 | XP_011539694.1 | |||
XM_011541393.2 | 2581 | UTR 3 | XP_011539695.1 | |||
XM_017001206.1 | 2581 | Intron | XP_016856695.1 | |||
XM_017001207.1 | 2581 | UTR 3 | XP_016856696.1 | |||
XM_017001208.1 | 2581 | Intron | XP_016856697.1 | |||
XM_017001209.1 | 2581 | UTR 3 | XP_016856698.1 |
MTF1 - metal regulatory transcription factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |