Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGCCTTTGTTCAGATTACATTTA[C/T]CATGTGGACCACCCATCACCTGGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604500 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO19 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MYO19 - myosin XIX | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033580.2 | Intron | NP_001028752.1 | ||||
NM_001163735.1 | Intron | NP_001157207.1 | ||||
NM_025109.5 | Intron | NP_079385.2 | ||||
XM_011525286.1 | Intron | XP_011523588.1 | ||||
XM_011525287.1 | Intron | XP_011523589.1 | ||||
XM_011525290.2 | Intron | XP_011523592.1 | ||||
XM_017025157.1 | Intron | XP_016880646.1 | ||||
XM_017025158.1 | Intron | XP_016880647.1 | ||||
XM_017025159.1 | Intron | XP_016880648.1 | ||||
XM_017025160.1 | Intron | XP_016880649.1 | ||||
XM_017025161.1 | Intron | XP_016880650.1 |
ZNHIT3 - zinc finger HIT-type containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |