Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATGGTATTAAAATGAGCAGGGA[A/G]TTTTTATGTTTGTGTCTCATCTTGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601513 | |||||||||||||||||||||||
Literature Links: |
FGF12 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
FGF12 - fibroblast growth factor 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004113.5 | 4320 | UTR 3 | NP_004104.3 | |||
NM_021032.4 | 4320 | UTR 3 | NP_066360.1 | |||
XM_005247227.2 | 4320 | UTR 3 | XP_005247284.1 | |||
XM_006713538.3 | 4320 | UTR 3 | XP_006713601.1 | |||
XM_017005879.1 | 4320 | UTR 3 | XP_016861368.1 |