Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATATAAAGAAATGACAACAGTAC[A/G]CAAGTGGTAACTTGTATAAAAGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613196 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ETAA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ETAA1 - Ewing tumor associated antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019002.3 | Intron | NP_061875.2 | ||||
XM_017004376.1 | Intron | XP_016859865.1 | ||||
XM_017004377.1 | Intron | XP_016859866.1 |