Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIML2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TRIML2 - tripartite motif family like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303419.1 | 1407 | Silent Mutation | GGC,GGG | G,G 396 | NP_001290348.1 | |
NM_173553.2 | 1407 | UTR 3 | NP_775824.2 | |||
XM_011531737.2 | 1407 | Silent Mutation | GGC,GGG | G,G 371 | XP_011530039.1 | |
XM_011531738.1 | 1407 | Silent Mutation | GGC,GGG | G,G 371 | XP_011530040.1 | |
XM_011531739.1 | 1407 | Silent Mutation | GGC,GGG | G,G 279 | XP_011530041.1 | |
XM_017007882.1 | 1407 | Silent Mutation | GGC,GGG | G,G 371 | XP_016863371.1 |
Set Membership: |
HapMap Validated |