Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
68 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608009 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AK5 - adenylate kinase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZZZ3 - zinc finger ZZ-type containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308237.1 | Intron | NP_001295166.1 | ||||
NM_015534.5 | Intron | NP_056349.1 | ||||
XM_005270725.3 | Intron | XP_005270782.1 | ||||
XM_005270726.3 | Intron | XP_005270783.1 | ||||
XM_005270727.3 | Intron | XP_005270784.1 | ||||
XM_005270729.4 | Intron | XP_005270786.1 | ||||
XM_017000970.1 | Intron | XP_016856459.1 | ||||
XM_017000971.1 | Intron | XP_016856460.1 | ||||
XM_017000972.1 | Intron | XP_016856461.1 |
Set Membership: |
HapMap Validated |