Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACGTCTTCATTAAAGTTTCCTGG[A/G]CAGAGTTTATTGTAACTAAGATCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603256 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRFIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRFIP1 - LRR binding FLII interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001137550.1 | Intron | NP_001131022.1 | ||||
NM_001137551.1 | Intron | NP_001131023.1 | ||||
NM_001137552.1 | Intron | NP_001131024.1 | ||||
NM_001137553.1 | Intron | NP_001131025.1 | ||||
NM_004735.3 | Intron | NP_004726.2 | ||||
XM_005246141.4 | Intron | XP_005246198.1 | ||||
XM_005246142.2 | Intron | XP_005246199.1 | ||||
XM_017005253.1 | Intron | XP_016860742.1 | ||||
XM_017005254.1 | Intron | XP_016860743.1 | ||||
XM_017005255.1 | Intron | XP_016860744.1 | ||||
XM_017005256.1 | Intron | XP_016860745.1 | ||||
XM_017005257.1 | Intron | XP_016860746.1 | ||||
XM_017005258.1 | Intron | XP_016860747.1 | ||||
XM_017005259.1 | Intron | XP_016860748.1 | ||||
XM_017005260.1 | Intron | XP_016860749.1 | ||||
XM_017005261.1 | Intron | XP_016860750.1 | ||||
XM_017005262.1 | Intron | XP_016860751.1 | ||||
XM_017005263.1 | Intron | XP_016860752.1 |