Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCAGGATCAGTGATTGAAATAA[C/T]TCAGAGTTAAAACAAAATGAATTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM44 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
RBM44 - RNA binding motif protein 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080504.2 | Intron | NP_001073973.2 | ||||
XM_005246075.1 | Intron | XP_005246132.1 | ||||
XM_011511152.2 | Intron | XP_011509454.1 | ||||
XM_011511153.2 | Intron | XP_011509455.1 | ||||
XM_011511154.2 | Intron | XP_011509456.1 | ||||
XM_011511155.1 | Intron | XP_011509457.1 | ||||
XM_017004054.1 | Intron | XP_016859543.1 | ||||
XM_017004055.1 | Intron | XP_016859544.1 | ||||
XM_017004056.1 | Intron | XP_016859545.1 | ||||
XM_017004057.1 | Intron | XP_016859546.1 | ||||
XM_017004058.1 | Intron | XP_016859547.1 |
Set Membership: |
HapMap Validated |