Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTTGACCCAGGTGTTGTTGCTG[C/T]TCCTTCCCCACTCCCCAATATATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616015 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF180 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF180 - ring finger protein 180 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113561.2 | Intron | NP_001107033.1 | ||||
NM_001323291.1 | Intron | NP_001310220.1 | ||||
NM_001323292.1 | Intron | NP_001310221.1 | ||||
NM_178532.4 | Intron | NP_848627.1 | ||||
XM_017009383.1 | Intron | XP_016864872.1 | ||||
XM_017009384.1 | Intron | XP_016864873.1 | ||||
XM_017009385.1 | Intron | XP_016864874.1 | ||||
XM_017009386.1 | Intron | XP_016864875.1 | ||||
XM_017009387.1 | Intron | XP_016864876.1 | ||||
XM_017009388.1 | Intron | XP_016864877.1 | ||||
XM_017009389.1 | Intron | XP_016864878.1 |