Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTCTGTAAGTATGTGGCCAAGAA[A/G]GGTACAGCAATTAATCATGCTTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615998 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RNF10 - ring finger protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014868.4 | Intron | NP_055683.3 | ||||
XM_005254013.2 | Intron | XP_005254070.1 | ||||
XM_006719717.2 | Intron | XP_006719780.1 | ||||
XM_017020282.1 | Intron | XP_016875771.1 | ||||
XM_017020283.1 | Intron | XP_016875772.1 |