Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
|||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607562 | |||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf141 PubMed Links | |||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU)
|
||||||
EAS - Not Available | African American
|
YRI (Yoruba)
|
||||||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
||||||
EUR - Not Available | ||||||||
AMR - Not Available |
C1orf141 - chromosome 1 open reading frame 141 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276351.1 | 382 | Intron | NP_001263280.1 | |||
NM_001276352.1 | 382 | Intron | NP_001263281.1 | |||
XM_011541465.2 | 382 | Intron | XP_011539767.1 | |||
XM_011541466.2 | 382 | Intron | XP_011539768.1 | |||
XM_011541467.1 | 382 | Intron | XP_011539769.1 | |||
XM_011541469.1 | 382 | Intron | XP_011539771.1 | |||
XM_011541472.1 | 382 | Intron | XP_011539774.1 | |||
XM_011541473.2 | 382 | Intron | XP_011539775.1 | |||
XM_011541475.1 | 382 | Intron | XP_011539777.1 | |||
XM_017001276.1 | 382 | Intron | XP_016856765.1 | |||
XM_017001277.1 | 382 | Intron | XP_016856766.1 |
IL23R - interleukin 23 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144701.2 | 382 | Intron | NP_653302.2 | |||
XM_005270516.2 | 382 | Intron | XP_005270573.1 | |||
XM_011540790.2 | 382 | UTR 5 | XP_011539092.1 | |||
XM_011540791.2 | 382 | Intron | XP_011539093.1 |
Set Membership: |
HapMap Validated |