Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCCAGTGCCTTGCGGGAACCCC[A/G]GCCTCATGACCAACCTCGGCCAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606811 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH4A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALDH4A1 - aldehyde dehydrogenase 4 family member A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IFFO2 - intermediate filament family orphan 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136265.1 | 2435 | UTR 3 | NP_001129737.1 | |||
XM_011540630.2 | 2435 | UTR 3 | XP_011538932.1 |