Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF345 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
ZNF345 - zinc finger protein 345 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242472.1 | 5026 | Intron | NP_001229401.1 | |||
NM_001242474.1 | 5026 | Intron | NP_001229403.1 | |||
NM_001242475.1 | 5026 | Intron | NP_001229404.1 | |||
NM_001242476.1 | 5026 | Intron | NP_001229405.1 | |||
NM_003419.4 | 5026 | Intron | NP_003410.1 | |||
XM_011526704.2 | 5026 | Intron | XP_011525006.1 | |||
XM_017026568.1 | 5026 | Intron | XP_016882057.1 | |||
XM_017026569.1 | 5026 | Intron | XP_016882058.1 | |||
XM_017026570.1 | 5026 | Intron | XP_016882059.1 | |||
XM_017026571.1 | 5026 | Intron | XP_016882060.1 | |||
XM_017026572.1 | 5026 | Intron | XP_016882061.1 | |||
XM_017026573.1 | 5026 | Intron | XP_016882062.1 |
ZNF829 - zinc finger protein 829 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037232.3 | 5026 | UTR 3 | NP_001032309.2 | |||
NM_001171979.1 | 5026 | UTR 3 | NP_001165450.1 | |||
XM_005258876.4 | 5026 | UTR 3 | XP_005258933.1 | |||
XM_011526933.2 | 5026 | UTR 3 | XP_011525235.1 |
Set Membership: |
HapMap Validated |