Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176947 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZAP70 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZAP70 - zeta chain of T cell receptor associated protein kinase 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079.3 | Intron | NP_001070.2 | ||||
NM_207519.1 | Intron | NP_997402.1 | ||||
XM_017004867.1 | Intron | XP_016860356.1 | ||||
XM_017004868.1 | Intron | XP_016860357.1 | ||||
XM_017004869.1 | Intron | XP_016860358.1 | ||||
XM_017004870.1 | Intron | XP_016860359.1 |
Set Membership: |
HapMap |