Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCTAGGTAACTTCTAAATGAGGC[A/G]TAGGTCCCACCATGCACCTGGATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARP8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PARP8 - poly(ADP-ribose) polymerase family member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178055.1 | Intron | NP_001171526.1 | ||||
NM_001178056.1 | Intron | NP_001171527.1 | ||||
NM_024615.3 | Intron | NP_078891.2 | ||||
XM_005248596.4 | Intron | XP_005248653.1 | ||||
XM_011543631.2 | Intron | XP_011541933.1 | ||||
XM_011543632.2 | Intron | XP_011541934.2 | ||||
XM_011543633.2 | Intron | XP_011541935.2 | ||||
XM_011543634.2 | Intron | XP_011541936.1 | ||||
XM_011543635.2 | Intron | XP_011541937.1 | ||||
XM_011543636.2 | Intron | XP_011541938.1 | ||||
XM_011543640.2 | Intron | XP_011541942.1 | ||||
XM_011543641.2 | Intron | XP_011541943.1 | ||||
XM_011543642.2 | Intron | XP_011541944.1 | ||||
XM_011543643.2 | Intron | XP_011541945.1 | ||||
XM_017009850.1 | Intron | XP_016865339.1 | ||||
XM_017009851.1 | Intron | XP_016865340.1 |