Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603607 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPS6KA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
RPS6KA5 - ribosomal protein S6 kinase A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322227.1 | Intron | NP_001309156.1 | ||||
NM_001322228.1 | Intron | NP_001309157.1 | ||||
NM_001322229.1 | Intron | NP_001309158.1 | ||||
NM_001322230.1 | Intron | NP_001309159.1 | ||||
NM_001322231.1 | Intron | NP_001309160.1 | ||||
NM_001322232.1 | Intron | NP_001309161.1 | ||||
NM_001322233.1 | Intron | NP_001309162.1 | ||||
NM_001322234.1 | Intron | NP_001309163.1 | ||||
NM_001322235.1 | Intron | NP_001309164.1 | ||||
NM_001322236.1 | Intron | NP_001309165.1 | ||||
NM_001322237.1 | Intron | NP_001309166.1 | ||||
NM_001322238.1 | Intron | NP_001309167.1 | ||||
NM_004755.3 | Intron | NP_004746.2 | ||||
NM_182398.2 | Intron | NP_872198.1 | ||||
XM_017021786.1 | Intron | XP_016877275.1 |
Set Membership: |
HapMap Validated |