Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615259 MIM: 190470 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
METTL21C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
METTL21C - methyltransferase like 21C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010977.2 | 348 | Silent Mutation | CTG,TTG | L,L 109 | NP_001010977.1 | |
XM_017020407.1 | 348 | Silent Mutation | CTG,TTG | L,L 109 | XP_016875896.1 | |
XM_017020408.1 | 348 | Silent Mutation | CTG,TTG | L,L 109 | XP_016875897.1 |
TPP2 - tripeptidyl peptidase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |