Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCCAAAACGCTGGAGCTCAGAA[C/G]CCAGCTCAGGTGCGCTCGTCGGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF26 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARHGEF26 - Rho guanine nucleotide exchange factor 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251962.1 | 224 | UTR 5 | NP_001238891.1 | |||
NM_001251963.1 | 224 | Intron | NP_001238892.1 | |||
NM_015595.3 | 224 | Intron | NP_056410.3 | |||
XM_011512672.1 | 224 | Intron | XP_011510974.1 | |||
XM_017006160.1 | 224 | Intron | XP_016861649.1 |
ARHGEF26-AS1 - ARHGEF26 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |