Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601870 MIM: 610993 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
METAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
METAP2 - methionyl aminopeptidase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP44 - ubiquitin specific peptidase 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042403.2 | Intron | NP_001035862.1 | ||||
NM_001278393.1 | Intron | NP_001265322.1 | ||||
NM_032147.4 | Intron | NP_115523.2 | ||||
XM_005269172.1 | Intron | XP_005269229.1 | ||||
XM_005269173.1 | Intron | XP_005269230.1 | ||||
XM_005269174.1 | Intron | XP_005269231.1 | ||||
XM_011538800.2 | Intron | XP_011537102.1 | ||||
XM_011538801.1 | Intron | XP_011537103.1 | ||||
XM_011538802.1 | Intron | XP_011537104.1 | ||||
XM_011538803.1 | Intron | XP_011537105.1 | ||||
XM_011538804.2 | Intron | XP_011537106.1 | ||||
XM_011538805.1 | Intron | XP_011537107.1 | ||||
XM_011538806.2 | Intron | XP_011537108.1 | ||||
XM_017020009.1 | Intron | XP_016875498.1 | ||||
XM_017020010.1 | Intron | XP_016875499.1 | ||||
XM_017020011.1 | Intron | XP_016875500.1 | ||||
XM_017020012.1 | Intron | XP_016875501.1 |
Set Membership: |
HapMap Validated |