Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGGCCATGGGGCTTTAACCTCT[C/G]CCTATATCTATAGGTTTTCTTTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601611 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC14A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC14A2 - solute carrier family 14 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242692.1 | Intron | NP_001229621.1 | ||||
NM_007163.3 | Intron | NP_009094.3 | ||||
XM_017026015.1 | Intron | XP_016881504.1 | ||||
XM_017026016.1 | Intron | XP_016881505.1 |