Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM68 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TMEM68 - transmembrane protein 68 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286657.1 | Intron | NP_001273586.1 | ||||
NM_001286660.1 | Intron | NP_001273589.1 | ||||
NM_001286661.1 | Intron | NP_001273590.1 | ||||
NM_152417.2 | Intron | NP_689630.1 | ||||
XM_005251150.1 | Intron | XP_005251207.1 | ||||
XM_011517454.1 | Intron | XP_011515756.1 | ||||
XM_011517455.2 | Intron | XP_011515757.1 | ||||
XM_017013040.1 | Intron | XP_016868529.1 |