Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGGACAGATGAAATTCAGCAGA[A/G]AGCCCTCTTTAGGTTACACCCTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 139311 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DKFZP434H168 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DKFZP434H168 - uncharacterized LOC26077 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GNAO1 - G protein subunit alpha o1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020988.2 | Intron | NP_066268.1 | ||||
NM_138736.2 | Intron | NP_620073.2 | ||||
XM_011523003.2 | Intron | XP_011521305.1 |
LOC283856 - uncharacterized LOC283856 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |