Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTGTAGCCTTCCTTTGCTTAAAA[A/C]CTTCTGAAAGTCAAACTCCTCACCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601568 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADD3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ADD3 - adducin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001121.3 | Intron | NP_001112.2 | ||||
NM_001320591.1 | Intron | NP_001307520.1 | ||||
NM_001320592.1 | Intron | NP_001307521.1 | ||||
NM_001320593.1 | Intron | NP_001307522.1 | ||||
NM_001320594.1 | Intron | NP_001307523.1 | ||||
NM_016824.4 | Intron | NP_058432.1 | ||||
NM_019903.4 | Intron | NP_063968.1 |
ADD3-AS1 - ADD3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |