Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607651 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHB1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PLEKHB1 - pleckstrin homology domain containing B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130033.1 | 739 | Intron | NP_001123505.1 | |||
NM_001130034.1 | 739 | Intron | NP_001123506.1 | |||
NM_001130035.1 | 739 | Intron | NP_001123507.1 | |||
NM_001130036.1 | 739 | Intron | NP_001123508.1 | |||
NM_021200.2 | 739 | Intron | NP_067023.1 | |||
XM_011545192.2 | 739 | UTR 5 | XP_011543494.1 | |||
XM_011545193.2 | 739 | UTR 5 | XP_011543495.1 |