Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCAAGCCCATTCTTACCAAGAAC[C/T]AATGAGTAAAATCAGGGGCCTCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM71F2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FAM71F2 - family with sequence similarity 71 member F2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012454.5 | 131 | UTR 5 | NP_001012457.3 | |||
NM_001128926.3 | 131 | UTR 5 | NP_001122398.1 | |||
NM_001290254.1 | 131 | UTR 5 | NP_001277183.1 | |||
NM_001290255.1 | 131 | UTR 5 | NP_001277184.1 | |||
XM_011516128.2 | 131 | UTR 5 | XP_011514430.1 | |||
XM_011516129.2 | 131 | UTR 5 | XP_011514431.1 | |||
XM_011516130.2 | 131 | UTR 5 | XP_011514432.1 | |||
XM_011516131.2 | 131 | UTR 5 | XP_011514433.1 | |||
XM_011516132.2 | 131 | UTR 5 | XP_011514434.1 | |||
XM_011516134.2 | 131 | Intron | XP_011514436.1 | |||
XM_011516135.2 | 131 | Intron | XP_011514437.1 | |||
XM_011516136.2 | 131 | Intron | XP_011514438.1 | |||
XM_017012147.1 | 131 | UTR 5 | XP_016867636.1 | |||
XM_017012149.1 | 131 | UTR 5 | XP_016867638.1 |