Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTACATTTAAGTCTGTAATTCA[A/G]TCGAGTTAATTTTTTGTGTATATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604731 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
USP15 - ubiquitin specific peptidase 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252078.1 | Intron | NP_001239007.1 | ||||
NM_001252079.1 | Intron | NP_001239008.1 | ||||
NM_006313.2 | Intron | NP_006304.1 | ||||
XM_005269261.2 | Intron | XP_005269318.1 | ||||
XM_006719718.3 | Intron | XP_006719781.1 | ||||
XM_017020284.1 | Intron | XP_016875773.1 | ||||
XM_017020285.1 | Intron | XP_016875774.1 |
Set Membership: |
HapMap |