Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605239 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP6V0A4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATP6V0A4 - ATPase H+ transporting V0 subunit a4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020632.2 | Intron | NP_065683.2 | ||||
NM_130840.2 | Intron | NP_570855.2 | ||||
NM_130841.2 | Intron | NP_570856.2 | ||||
XM_005250393.1 | Intron | XP_005250450.1 | ||||
XM_005250394.3 | Intron | XP_005250451.1 |
TMEM213 - transmembrane protein 213 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085429.1 | Intron | NP_001078898.1 | ||||
XM_005250167.3 | Intron | XP_005250224.1 |
Set Membership: |
HapMap |