Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGCCAGACCTTTGGCCAAATCC[A/T]GATGGTGTCTCCAGCAGGGCAGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602291 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXJ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FOXJ1 - forkhead box J1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF157 - ring finger protein 157 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052916.2 | Intron | NP_443148.1 | ||||
XM_005257007.4 | Intron | XP_005257064.1 | ||||
XM_011524273.2 | Intron | XP_011522575.1 | ||||
XM_017024117.1 | Intron | XP_016879606.1 | ||||
XM_017024118.1 | Intron | XP_016879607.1 | ||||
XM_017024119.1 | Intron | XP_016879608.1 | ||||
XM_017024120.1 | Intron | XP_016879609.1 |
RNF157-AS1 - RNF157 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |