Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGTGATAATCAGAAAATTCCCTT[C/T]CTCTGTATTCATGAACAGAAGTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609248 MIM: 604479 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HERC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HERC4 - HECT and RLD domain containing E3 ubiquitin protein ligase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278185.1 | Intron | NP_001265114.1 | ||||
NM_001278186.1 | Intron | NP_001265115.1 | ||||
NM_001278187.1 | Intron | NP_001265116.1 | ||||
NM_015601.3 | Intron | NP_056416.2 | ||||
NM_022079.2 | Intron | NP_071362.1 | ||||
XM_011539592.2 | Intron | XP_011537894.1 | ||||
XM_011539593.2 | Intron | XP_011537895.1 | ||||
XM_011539594.2 | Intron | XP_011537896.1 | ||||
XM_011539595.2 | Intron | XP_011537897.1 | ||||
XM_011539596.2 | Intron | XP_011537898.1 | ||||
XM_011539597.2 | Intron | XP_011537899.1 | ||||
XM_017016041.1 | Intron | XP_016871530.1 |
SIRT1 - sirtuin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |