Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTCCATTGGTGCCCTGGGTGGC[G/T]TGGGATCAATGACATCCATTCCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTBD16 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BTBD16 - BTB domain containing 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318189.1 | Intron | NP_001305118.1 | ||||
NM_144587.3 | Intron | NP_653188.2 | ||||
XM_011539239.2 | Intron | XP_011537541.1 | ||||
XM_011539240.2 | Intron | XP_011537542.1 | ||||
XM_011539241.2 | Intron | XP_011537543.1 | ||||
XM_011539242.2 | Intron | XP_011537544.1 | ||||
XM_017015636.1 | Intron | XP_016871125.1 | ||||
XM_017015637.1 | Intron | XP_016871126.1 |