Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGATCTGAGATGGGAAGACAGGAA[A/T]GGTAATCTCACATCTCCATCCTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611752 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf40 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2orf40 - chromosome 2 open reading frame 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032411.2 | Intron | NP_115787.1 | ||||
XM_006712799.3 | Intron | XP_006712862.1 | ||||
XM_011511993.2 | Intron | XP_011510295.1 | ||||
XM_017005106.1 | Intron | XP_016860595.1 |