Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605014 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STX11 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
STX11 - syntaxin 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003764.3 | Intron | NP_003755.2 | ||||
XM_011536213.2 | Intron | XP_011534515.1 | ||||
XM_011536214.2 | Intron | XP_011534516.1 | ||||
XM_011536217.2 | Intron | XP_011534519.1 | ||||
XM_011536218.2 | Intron | XP_011534520.1 | ||||
XM_017011400.1 | Intron | XP_016866889.1 |
Set Membership: |
HapMap Validated |