Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCGTGCAGACTGGATAAAGGCTG[A/G]AGGAGAAGAGTTTTGGAAGGGCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604122 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIGB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PIGB - phosphatidylinositol glycan anchor biosynthesis class B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004855.4 | Intron | NP_004846.4 | ||||
XM_005254795.4 | Intron | XP_005254852.1 | ||||
XM_011522235.2 | Intron | XP_011520537.1 | ||||
XM_011522236.2 | Intron | XP_011520538.1 | ||||
XM_011522237.2 | Intron | XP_011520539.1 | ||||
XM_017022730.1 | Intron | XP_016878219.1 | ||||
XM_017022731.1 | Intron | XP_016878220.1 | ||||
XM_017022732.1 | Intron | XP_016878221.1 |
Set Membership: |
HapMap |