Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTATCATTTTCAACAGGAAGAACC[A/C]TGCCAAATGTTGTTTGTCTGATCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607837 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLN8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CLN8 - ceroid-lipofuscinosis, neuronal 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018941.3 | Intron | NP_061764.2 | ||||
XM_005266021.3 | Intron | XP_005266078.1 | ||||
XM_005266022.1 | Intron | XP_005266079.1 | ||||
XM_005266023.1 | Intron | XP_005266080.1 | ||||
XM_011534745.1 | Intron | XP_011533047.1 | ||||
XM_011534746.2 | Intron | XP_011533048.1 | ||||
XM_011534747.1 | Intron | XP_011533049.1 |
LOC101927752 - uncharacterized LOC101927752 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |