Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGGGTGCACGTGGAAGGCGGAG[A/T]GGAGCCCAACAGCCAGCGGCTGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 156352 MIM: 156353 MIM: 156354 MIM: 156355 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MT1F PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MT1F - metallothionein 1F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MT1G - metallothionein 1G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301267.1 | Intron | NP_001288196.1 | ||||
NM_005950.2 | Intron | NP_005941.1 |
MT1H - metallothionein 1H | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MT1IP - metallothionein 1I, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |