Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATTATTTAGGCCGATGAGTAGGA[C/T]GGTCAAGGAATTTATAATTTCCAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF618 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF618 - zinc finger protein 618 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318040.1 | Intron | NP_001304969.1 | ||||
NM_001318041.1 | Intron | NP_001304970.1 | ||||
NM_001318042.1 | Intron | NP_001304971.1 | ||||
NM_133374.2 | Intron | NP_588615.2 | ||||
XM_005251692.2 | Intron | XP_005251749.1 | ||||
XM_005251699.2 | Intron | XP_005251756.1 | ||||
XM_011518200.1 | Intron | XP_011516502.1 | ||||
XM_011518201.1 | Intron | XP_011516503.1 | ||||
XM_011518203.2 | Intron | XP_011516505.1 | ||||
XM_011518204.2 | Intron | XP_011516506.1 | ||||
XM_011518209.2 | Intron | XP_011516511.1 | ||||
XM_017014241.1 | Intron | XP_016869730.1 | ||||
XM_017014242.1 | Intron | XP_016869731.1 | ||||
XM_017014243.1 | Intron | XP_016869732.1 | ||||
XM_017014244.1 | Intron | XP_016869733.1 | ||||
XM_017014245.1 | Intron | XP_016869734.1 | ||||
XM_017014246.1 | Intron | XP_016869735.1 | ||||
XM_017014247.1 | Intron | XP_016869736.1 | ||||
XM_017014248.1 | Intron | XP_016869737.1 | ||||
XM_017014249.1 | Intron | XP_016869738.1 | ||||
XM_017014250.1 | Intron | XP_016869739.1 | ||||
XM_017014251.1 | Intron | XP_016869740.1 | ||||
XM_017014252.1 | Intron | XP_016869741.1 |
Set Membership: |
HapMap |