Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACACACTAAGACTTTAGCCCTAA[C/T]CTCAGGCCCAGTTCTGGCTAGGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602630 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TGIF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TGIF1 - TGFB induced factor homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278682.1 | Intron | NP_001265611.1 | ||||
NM_001278684.1 | Intron | NP_001265613.1 | ||||
NM_001278686.1 | Intron | NP_001265615.1 | ||||
NM_003244.3 | Intron | NP_003235.1 | ||||
NM_170695.3 | Intron | NP_733796.2 | ||||
NM_173207.2 | Intron | NP_775299.1 | ||||
NM_173208.2 | Intron | NP_775300.1 | ||||
NM_173209.2 | Intron | NP_775301.1 | ||||
NM_173210.2 | Intron | NP_775302.1 | ||||
NM_173211.1 | Intron | NP_775303.1 | ||||
NM_174886.2 | Intron | NP_777480.1 | ||||
XM_011525735.2 | Intron | XP_011524037.1 | ||||
XM_017025958.1 | Intron | XP_016881447.1 | ||||
XM_017025959.1 | Intron | XP_016881448.1 |