Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAACTGACCTATTTTAAACAAGAT[G/T]GCTGTAGTGAATTCCAATTGAGTTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603297 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DYNC2H1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DYNC2H1 - dynein cytoplasmic 2 heavy chain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080463.1 | Intron | NP_001073932.1 | ||||
NM_001377.2 | Intron | NP_001368.2 | ||||
XM_006718903.2 | Intron | XP_006718966.1 | ||||
XM_017018291.1 | Intron | XP_016873780.1 | ||||
XM_017018292.1 | Intron | XP_016873781.1 | ||||
XM_017018293.1 | Intron | XP_016873782.1 |
Set Membership: |
HapMap |