Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATATATCTTATGATTCAAGTCA[G/T]AATTTAAATAGAATTGAAGATGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614315 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACOT12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ACOT12 - acyl-CoA thioesterase 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CKMT2-AS1 - CKMT2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZCCHC9 - zinc finger CCHC-type containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131035.1 | Intron | NP_001124507.1 | ||||
NM_001131036.1 | Intron | NP_001124508.1 | ||||
NM_032280.2 | Intron | NP_115656.1 | ||||
XM_005248616.3 | Intron | XP_005248673.1 |