Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF385D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF385D - zinc finger protein 385D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024697.2 | 1719 | Silent Mutation | TCC,TCT | S,S 369 | NP_078973.1 | |
XM_011534122.1 | 1719 | Intron | XP_011532424.1 | |||
XM_011534123.2 | 1719 | Intron | XP_011532425.1 | |||
XM_011534124.2 | 1719 | Intron | XP_011532426.1 | |||
XM_017007191.1 | 1719 | Silent Mutation | TCC,TCT | S,S 489 | XP_016862680.1 | |
XM_017007192.1 | 1719 | Silent Mutation | TCC,TCT | S,S 470 | XP_016862681.1 | |
XM_017007193.1 | 1719 | Silent Mutation | TCC,TCT | S,S 418 | XP_016862682.1 | |
XM_017007194.1 | 1719 | Silent Mutation | TCC,TCT | S,S 405 | XP_016862683.1 | |
XM_017007195.1 | 1719 | Silent Mutation | TCC,TCT | S,S 403 | XP_016862684.1 | |
XM_017007196.1 | 1719 | Silent Mutation | TCC,TCT | S,S 388 | XP_016862685.1 | |
XM_017007197.1 | 1719 | Silent Mutation | TCC,TCT | S,S 385 | XP_016862686.1 | |
XM_017007198.1 | 1719 | Silent Mutation | TCC,TCT | S,S 384 | XP_016862687.1 | |
XM_017007199.1 | 1719 | Silent Mutation | TCC,TCT | S,S 336 | XP_016862688.1 | |
XM_017007200.1 | 1719 | Silent Mutation | TCC,TCT | S,S 333 | XP_016862689.1 | |
XM_017007201.1 | 1719 | Silent Mutation | TCC,TCT | S,S 275 | XP_016862690.1 | |
XM_017007202.1 | 1719 | Silent Mutation | TCC,TCT | S,S 236 | XP_016862691.1 | |
XM_017007203.1 | 1719 | Intron | XP_016862692.1 |