Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGCATGCAGACCAGAGTAATGA[G/T]ATTTTTATGTTGACCGCAATGCGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZCCHC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZCCHC2 - zinc finger CCHC-type containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017742.5 | Intron | NP_060212.4 | ||||
XM_006722493.3 | Intron | XP_006722556.1 | ||||
XM_011526044.1 | Intron | XP_011524346.1 | ||||
XM_017025802.1 | Intron | XP_016881291.1 |
Set Membership: |
Validated |