Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATGGATGCCTTGGTAGTAAGATC[A/G]GAGGGCTGAATCTGGAGACTTGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605984 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EED PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EED - embryonic ectoderm development | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308007.1 | Intron | NP_001294936.1 | ||||
NM_003797.4 | Intron | NP_003788.2 | ||||
XM_005274373.2 | Intron | XP_005274430.1 | ||||
XM_011545330.1 | Intron | XP_011543632.1 | ||||
XM_011545331.1 | Intron | XP_011543633.1 | ||||
XM_017018512.1 | Intron | XP_016874001.1 | ||||
XM_017018513.1 | Intron | XP_016874002.1 |