Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAACATGAGGGATGGAGTAGATGA[T/C]GTTAGGGAGGGGGATGACAAGGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608124 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
XYLT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
XYLT1 - xylosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022166.3 | 7983 | UTR 3 | NP_071449.1 | |||
XM_017023539.1 | 7983 | Intron | XP_016879028.1 | |||
XM_017023540.1 | 7983 | Intron | XP_016879029.1 |
Set Membership: |
HapMap Validated |