Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGAGGAAGATGCTAGGAATAAAT[A/C]CCCCTTTTACACATGGGGAAACTGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARRDC5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARRDC5 - arrestin domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080523.1 | Intron | NP_001073992.1 | ||||
XM_005259616.2 | Intron | XP_005259673.1 | ||||
XM_011528181.2 | Intron | XP_011526483.1 | ||||
XM_017027144.1 | Intron | XP_016882633.1 | ||||
XM_017027145.1 | Intron | XP_016882634.1 |