Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCACACACAGCCACATGACACACA[A/C]GCCAAACCCTGACAGTGTTGCCCAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
49 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614264 MIM: 609607 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP30 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ARHGAP30 - Rho GTPase activating protein 30 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NECTIN4 - nectin cell adhesion molecule 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030916.2 | 2116 | UTR 3 | NP_112178.2 | |||
XM_005245508.3 | 2116 | UTR 3 | XP_005245565.1 | |||
XM_011510021.2 | 2116 | UTR 3 | XP_011508323.1 | |||
XM_011510022.2 | 2116 | UTR 3 | XP_011508324.1 | |||
XM_011510023.1 | 2116 | Intron | XP_011508325.1 |